Merck
  • Polyacetylated labdane-type diterpenoids, ptychantins P-R from Chinese liverwort Ptychanthus striatus.
Polyacetylated labdane-type diterpenoids, ptychantins P-R from Chinese liverwort Ptychanthus striatus.
Journal of Asian natural products research (2015-02-14)
Jing-Yi Wu, Jiao-Zhen Zhang, Ya-Qi Kang, Xiao Wang, Pei-Hong Fan, Jin-Chuan Zhou, Hong-Xiang Lou
ABSTRACT
Three new polyacetylated labdane diterpenoids ptychantins P-R (1-3) and four known compounds (4-7) were isolated from an EtOH extract of the Chinese liverwort Ptychanthus striatus (Lehm. & Lindenb.) Nees. Their structures were established by extensive analysis of spectroscopic data (IR, UV, HRESIMS, 1D NMR, and 2D NMR).
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous