Merck
  • Reference values of hair toxic trace elements content in occupationally non-exposed Russian population.
Reference values of hair toxic trace elements content in occupationally non-exposed Russian population.
Environmental toxicology and pharmacology (2015-06-10)
Anatoly V Skalny, Margarita G Skalnaya, Alexey A Tinkov, Eugeny P Serebryansky, Vasily A Demidov, Yulia N Lobanova, Andrei R Grabeklis, Elena S Berezkina, Irina V Gryazeva, Andrey A Skalny, Alexandr A Nikonorov
ABSTRACT
A total of 5908 occupationally non-exposed adults (4384 women and 1524 men) living in Moscow and Moscow region were involved in the current investigation. Hair Al, As, Be, Bi, Cd, Hg, Li, Ni, Pb, Sn, and Sr content was estimated by inductively-coupled plasma mass spectrometry using NexION 300D. Men are characterized by significantly higher hair Al, As, Cd, Hg, Li, and Pb content. At the same time, hair levels of Bi, Ni, Sn, and Sr were significantly higher in women. Consequently, the reference ranges were estimated for male, female, and general cohort as coverage intervals in accordance with IUPAC recommendations.
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder