Merck
  • Characterization and functional analysis of porcine estrogen-related receptors and their alternative splicing variants.
Characterization and functional analysis of porcine estrogen-related receptors and their alternative splicing variants.
Journal of animal science (2015-10-07)
F Yang, X Du, Y Wang, C Wang, C Huang, Q Xiao, X Bai, H Wang
ABSTRACT
Estrogen-related receptors (ESRR) are orphan nuclear hormone receptors with unidentified ligands; they play important roles in tissue regulation and development and maintenance of pluripotent cell identity. The splicer variant, genomic organization, and physiological roles of ESRR have been elucidated in the human and the mouse. However, in livestock, they remain elusive. In this study, we cloned porcine ESRR family members , , and . Two alternative splicing variants, and , and a novel were identified. To determine the domain function, we constructed vectors with sequential deletions of the ESRRB coding sequence. The functional analysis showed that the C domain of ESRR plays a core role in promoting the activation of estrogen response elements that are found in all kinds of ESRR-targeting genes, whereas the E domain is not essential for transcription regulation of ESRR unless a specific and identified ligand is applied.
Sigma-Aldrich
Sodium trichloroacetate, 97%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%