Merck
  • Extraction of rebaudioside-A by sonication from Stevia rebaudiana Bertoni leaf and decolorization of the extract by polymers.
Extraction of rebaudioside-A by sonication from Stevia rebaudiana Bertoni leaf and decolorization of the extract by polymers.
Journal of food science and technology (2015-09-08)
Mohammed Abdalbasit A Gasmalla, Ruijin Yang, Xiao Hua
ABSTRACT
Optimization of steviol glycosides extraction from Stevia rebaudiana Bertoni leaf was carried out by investigating the effects of isopropyl alcohol concentration (60 %, v/v), time (6-24 min), temperature (30 °C) and sonic power (300-480 W) on extraction of rebaudioside A from Stevia rebaudiana leaves and decolorization of the extract by polymer (Separan AP30 and Resin ADS-7). The results showed that isopropyl alcohol was suitable for the extraction of rebaudioside A from Stevia rebaudiana leaves and the yield of rebaudioside A achieved 35.61 g/100 g when the output power was 360 W and treatment time was 18 min. The sonication had influence on the particle size of stevia leaf and the color of the extracted solution. As the sonication intensity increased, the particle size decreased. The colour of differently treated stevia solutions were significantly different (P < 0.05). Separan AP30 and adsorption resin ADS-7 were performed to remove the colour impurity. The results showed that more than 65 % of the coloured impurity was removed by Separan AP30 combined with Calcium oxide (CaO).
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC