Merck
  • Use of LC-MS-MS as an alternative to currently available immunoassay methods to quantitate corticosterone in egg yolk and albumen.
Use of LC-MS-MS as an alternative to currently available immunoassay methods to quantitate corticosterone in egg yolk and albumen.
Analytical and bioanalytical chemistry (2014-11-20)
Siegrid De Baere, Tom Rosendahl Larsen, Mathias Devreese, Patrick De Backer, Liesbeth De Neve, Graham Fairhurst, Luc Lens, Siska Croubels
ABSTRACT
Corticosterone (CORT) is the dominant plasma glucocorticoid in birds. There has been increasing interest in the function of CORT in avian egg yolk and in the potential to use CORT concentrations in eggs to quantify stress and to assess the effect of maternal stress on offspring. The concentration of CORT in egg yolk is most frequently assessed using enzyme or radioimmunoassays, alone or in combination with high-performance liquid chromatography. However, the quantification of CORT is frequently hampered by the presence of high concentrations of other steroid hormones which cross-react with the CORT antibody. As an alternative, we developed a sensitive and specific LC-MS-MS method. The sample-preparation procedure consisted of a protein-lipid precipitation step, followed by defatting and clean-up using a C18 SPE column. Chromatography was performed on an Acquity C18 BEH column (50 mm × 2.1 mm i.d., dp: 1.7 μm, run-time: 6 min), using 0.1% formic acid in both water (A) and acetonitrile (B) as mobile phases. The MS-MS instrument was operated in the positive-electrospray-ionization mode. The method was validated in-house according to European Guidelines (linearity, accuracy and precision, limits of quantification and detection, specificity, stability) and the results fell within the accepted ranges. The method was successfully used for the analysis of CORT in yolk and albumen of eggs collected from eight breeding lesser black-backed gulls at a Flemish coastal colony. CORT concentrations were in the range 42.4-166.3 pg g(-1) in albumen and < LOQ (75 pg g(-1))-762.5 pg g(-1) in yolk.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
5-Pregnen-3β-ol-20-one, ≥98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate