Merck
  • Genomic DNA extraction methods using formalin-fixed paraffin-embedded tissue.
Genomic DNA extraction methods using formalin-fixed paraffin-embedded tissue.
Analytical biochemistry (2015-07-02)
Keerti Potluri, Ahmed Mahas, Michael N Kent, Sameep Naik, Michael Markey
ABSTRACT
As new technologies come within reach for the average cytogenetic laboratory, the study of chromosome structure has become increasingly more sophisticated. Resolution has improved from karyotyping (in which whole chromosomes are discernible) to fluorescence in situ hybridization and comparative genomic hybridization (CGH, with which specific megabase regions are visualized), array-based CGH (aCGH, examining hundreds of base pairs), and next-generation sequencing (providing single base pair resolution). Whole genome next-generation sequencing remains a cost-prohibitive method for many investigators. Meanwhile, the cost of aCGH has been reduced during recent years, even as resolution has increased and protocols have simplified. However, aCGH presents its own set of unique challenges. DNA of sufficient quantity and quality to hybridize to arrays and provide meaningful results is required. This is especially difficult for DNA from formalin-fixed paraffin-embedded (FFPE) tissues. Here, we compare three different methods for acquiring DNA of sufficient length, purity, and "amplifiability" for aCGH and other downstream applications. Phenol-chloroform extraction and column-based commercial kits were compared with adaptive focused acoustics (AFA). Of the three extraction methods, AFA samples showed increased amplicon length and decreased polymerase chain reaction (PCR) failure rate. These findings support AFA as an improvement over previous DNA extraction methods for FFPE tissues.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
3-Methylbutanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
3-Methylbutanol, BioReagent, ≥98.5%, Molecular Biology
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis