Merck
  • A novel polymeric monolith prepared with multi-acrylate crosslinker for retention-independent efficient separation of small molecules in capillary liquid chromatography.
A novel polymeric monolith prepared with multi-acrylate crosslinker for retention-independent efficient separation of small molecules in capillary liquid chromatography.
Analytica chimica acta (2015-06-20)
Haiyang Zhang, Junjie Ou, Yinmao Wei, Hongwei Wang, Zhongshan Liu, Lianfang Chen, Hanfa Zou
ABSTRACT
Low column efficiency for small molecules in reversed-phase chromatography is a major problem commonly encountered in polymer-based monoliths. Herein, a novel highly crosslinked porous polymeric monolith was in situ prepared by using a multi-acrylate monomer, dipentaerythritol penta-/hexa-acrylate (DPEPA), as crosslinker, which copolymerized with lauryl methacrylate (LMA) as functional monomer in a UV-transparent fused-silica capillary via photo-initiated free-radical polymerization within 5 min. The mechanical stability and permeability of the resulting poly(LMA-co-DPEPA) monolith were characterized in detail. One series of highly crosslinked poly(LMA-co-DPEPA) columns were prepared with relatively higher content of crosslinker (63.3%) in the precursor. Although they exhibited lower permeability, high column efficiency for alkylbenzenes was acquired in cLC, and the minimum plate height (column B) was in the range of 6.04-9.00 μm, corresponding to 111,000-165,000 N m(-1). Meanwhile, another series of poly(LMA-co-DPEPA) columns prepared with relatively lower content of crosslinker (52.7%) in the precursor exhibited higher permeability, but the minimum plate height (column E) was relatively low in the range of 10.75-20.04 μm for alkylbenzenes, corresponding to 50,000-93,000 N m(-1). Compared with common poly(LMA-co-EDMA) columns previously reported, the highly crosslinked poly(LMA-co-DPEPA) columns using a multi-acrylate monomer as crosslinker possessed remarkably high column efficiency for small molecules in cLC. By plotting of plate height (H) of alkylbenzenes versus the linear velocity (u) of mobile phase, the results revealed a retention-independent efficient performance of small molecules in the isocratic elution, indicating that the use of multi-functional crosslinker possibly prevents the generation of gel-like micropores in the poly(LMA-co-DPEPA) monolith, reducing the mass transfer resistance (C-term).
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Hexyl alcohol, FCC, FG
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
4-Methoxyphenol, ReagentPlus®, 99%
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
Hexyl alcohol, natural, ≥98%, FCC, FG