Merck
  • Inactivation of Protein Tyrosine Phosphatases by Peracids Correlates with the Hydrocarbon Chain Length.
Inactivation of Protein Tyrosine Phosphatases by Peracids Correlates with the Hydrocarbon Chain Length.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-06-27)
Alicja Kuban-Jankowska, Magdalena Gorska, Jack A Tuszynski, Cassandra D M Churchill, Philip Winter, Mariusz Klobukowski, Michal Wozniak
ABSTRACT
Protein tyrosine phosphatases are crucial enzymes controlling numerous physiological and pathophysiological events and can be regulated by oxidation of the catalytic domain cysteine residue. Peracids are highly oxidizing compounds, and thus may induce inactivation of PTPs. The aim of the present study was to evaluate the inhibitory effect of peracids with different length of hydrocarbon chain on the activity of selected PTPs. The enzymatic activity of human CD45, PTP1B, LAR, bacterial YopH was assayed under the cell-free conditions, and activity of cellular CD45 in human Jurkat cell lysates. The molecular docking and molecular dynamics were performed to evaluate the peracids binding to the CD45 active site. Here we demonstrate that peracids reduce enzymatic activity of recombinant CD45, PTP1B, LAR, YopH and cellular CD45. Our studies indicate that peracids are more potent inhibitors of CD45 than hydrogen peroxide (with an IC50 value equal to 25 nM for peroctanoic acid and 8 µM for hydrogen peroxide). The experimental data show that the inactivation caused by peracids is dependent on hydrocarbon chain length of peracids with maximum inhibitory effect of medium-chain peracids (C8-C12 acyl chain), which correlates with calculated binding affinities to the CD45 active site. Peracids are potent inhibitors of PTPs with the strongest inhibitory effect observed for medium-chain peracids.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
CD45 Active human, recombinant, expressed in baculovirus infected insect cells, ≥90% (SDS-PAGE)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
p-Terphenyl, ≥99.5% (HPLC)