Merck
  • Biodegradation of pharmaceuticals in hospital wastewater by staged Moving Bed Biofilm Reactors (MBBR).
Biodegradation of pharmaceuticals in hospital wastewater by staged Moving Bed Biofilm Reactors (MBBR).
Water research (2015-07-15)
Mònica Escolà Casas, Ravi Kumar Chhetri, Gordon Ooi, Kamilla M S Hansen, Klaus Litty, Magnus Christensson, Caroline Kragelund, Henrik R Andersen, Kai Bester
ABSTRACT
Hospital wastewater represents a significant input of pharmaceuticals into municipal wastewater. As Moving Bed Biofilm Reactors (MBBRs) appear to remove organic micro-pollutants, hospital wastewater was treated with a pilot plant consisting of three MBBRs in series. The removal of pharmaceuticals was studied in two experiments: 1) A batch experiment where pharmaceuticals were spiked to each reactor and 2) a continuous flow experiment at native concentrations. DOC removal, nitrification as well as removal of pharmaceuticals (including X-ray contrast media, β-blockers, analgesics and antibiotics) occurred mainly in the first reactor. In the batch experiment most of the compounds followed a single first-order kinetics degradation function, giving degradation rate constants ranged from 5.77 × 10(-3) to 4.07 h(-1), from -5.53 × 10(-3) to 9.24 × 10(-1) h(-1) and from 1.83 × 10(-3) to 2.42 × 10(-1) h(-1) for first, second and third reactor respectively. Generally, the highest removal rate constants were found in the first reactor while the lowest were found in the third one. This order was inverted for most compounds, when the removal rate constants were normalized to biomass, indicating that the last tank had the most effective biofilms. In the batch experiment, 21 out of 26 compounds were assessed to be degraded with more than 20% within the MBBR train. In the continuous flow experiment the measured removal rates were lower than those estimated from the batch experiments.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Diatrizoic acid, Iodine-containing contrast agent
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Supelco
Ibuprofen