Merck
Th17 cytokines regulate osteoclastogenesis in rheumatoid arthritis.
The American journal of pathology (2015-09-13)
Kyoung-Woon Kim, Hae-Rim Kim, Bo-Mi Kim, Mi-La Cho, Sang-Heon Lee
ABSTRACT
This study determined the effect of type 17 helper T-cell (Th17) cytokines on osteoclastogenesis in rheumatoid arthritis (RA). The expression of IL-17 and receptor activator of NF-κB ligand (RANKL) was determined in synovial tissue, fibroblast-like synoviocytes (FLSs), and synovial fluids of RA patients using immunostaining and enzyme-linked immunosorbent assay. Th17 cytokine-induced RANKL expression was studied in RA FLS by using real-time PCR, luciferase activity assays, and Western blot analysis. Human peripheral blood monocytes were cultured with macrophage colony-stimulating factor and Th17 cytokines, after which osteoclastogenesis was evaluated by counting the number of tartrate-resistant acid phosphatase-positive multinucleated cells. Osteoclastogenesis was also evaluated after monocytes were co-cultured with IL-17-prestimulated FLS. There was significant correlation between RANKL and IL-17 levels in RA synovial fluid. IL-17, IL-21, and IL-22 increased the expression of Rankl mRNA in RA FLS, and the IL-17-induced RANKL expression decreased by the inhibition of Act1, tumor necrosis factor receptor-associated factor 6, NF-κB, and activator protein-1. Th17 cytokines and IL-17-prestimulated FLS induced osteoclastogenesis from monocytes in the absence of exogenous RANKL. The osteoclastic effect was reduced by inhibition of tumor necrosis factor-α. Th17 cytokines have a dual effect on osteoclastogenesis in RA: direct induction of osteoclastogenesis from monocytes and up-regulation of RANKL production in RA FLS. This Th17 cytokine/RANKL axis could be a potential therapeutic target for bone destruction in RA.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
IL-17 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Interleukin-17A human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Interleukin-17A human, IL-17A, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Parthenolide, ≥98% (HPLC)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%