Merck
  • Half-life of DISC1 protein and its pathological significance under hypoxia stress.
Half-life of DISC1 protein and its pathological significance under hypoxia stress.
Neuroscience research (2015-03-05)
Sandeep Kumar Barodia, Sang Ki Park, Koko Ishizuka, Akira Sawa, Atsushi Kamiya
ABSTRACT
DISC1 (disrupted in schizophrenia 1) is an intracellular scaffolding molecule which regulates multiple signaling pathways for neural cell differentiation and function. Many biological studies utilizing animal models of DISC1 have indicated that loss of DISC1 functions are associated with pathological psychiatric conditions. Thus, DISC1 protein stability is a prerequisite to its goal in governing neural function, and modulating the protein stability of DISC1 may be a key target for understanding underlying pathology, as well promising drug discovery strategies. Nonetheless, a half-life of DISC1 protein has remained unexplored. Here, we determine for the first time the half-life of DISC1, which are regulated by ubiquitin-proteasome cascade. Overexpression of PDE4B2, a binding partner of DISC1, prolonged the half-life of DISC1, whereas NDEL1 does not alter DISC1 protein stability. Notably, the half-life of DISC1 is diminished under hypoxia stress by increasing protein degradation of DISC1, suggesting that alteration of DISC1 stability may be involved in hypoxia stress-mediated pathological conditions, such as ischemic stroke.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
SAFC
L-Methionine
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%