Merck
  • A rapid LC-MS/MS method for quantitative profiling of fatty acids, sterols, glycerolipids, glycerophospholipids and sphingolipids in grapes.
A rapid LC-MS/MS method for quantitative profiling of fatty acids, sterols, glycerolipids, glycerophospholipids and sphingolipids in grapes.
Talanta (2015-06-07)
Anna Della Corte, Giulia Chitarrini, Iole Maria Di Gangi, Domenico Masuero, Evelyn Soini, Fulvio Mattivi, Urska Vrhovsek
ABSTRACT
The abundance of lipids in plants is influenced by genotype and phenotype. Despite being a very important class of plant metabolites, knowledge of grape lipids is still very limited to date, with the exception of those located in seeds. Few investigations of grape lipids have shown that their profile depends on grape maturity, the variety and their location in the berry. Recent advances in liquid chromatography coupled to mass spectrometry have paved the way for faster analysis of lipids with minimal sample preparation. Here we describe a validation method for the extraction, identification and quantification of different classes of grape lipids: fatty acids, sterols, glycerolipids, glycerophospholipids and sphingolipids using liquid chromatographic electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS). The method was validated for 33 lipids, with linearity range (R(2)=0.95-1.00), LOQ (0.003-14.88 ng mL(-1)) and intraday and interday repeatability being evaluated for each lipid. The lipid profiling method developed was successfully applied to the analysis of 18 grape samples (10 red grape and 8 white grape varieties) from 4 different genetic groups: Vitis vinifera, Vitis non-vinifera, Muscat and hybrid; 33 lipids were identified and quantified. This method, which can be easily expanded to include further compounds and other plant tissues, is the starting point for analysis of the lipid profile in different grape tissues, an essential goal for better understanding the role of lipids in grape physiology.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis