Merck
  • Size reduction of cosolvent-infused microbubbles to form acoustically responsive monodisperse perfluorocarbon nanodroplets.
Size reduction of cosolvent-infused microbubbles to form acoustically responsive monodisperse perfluorocarbon nanodroplets.
Lab on a chip (2015-07-30)
Minseok Seo, Ross Williams, Naomi Matsuura
ABSTRACT
Perfluorocarbon (PFC) nanodroplet agents are exciting new biomaterials that can be remotely vapourized by ultrasound or light to change into micron-scale gas bubbles in situ. After PFC nanodroplet vapourization, the micron-scale gas bubble can interact strongly with ultrasound radiation, such that the bubbles can be used for cancer imaging and therapy. For these phase-change agents to be useful, however, PFC nanodroplets must be produced in the range of 100 to 400 nm in diameter with high size control and monodispersity, restrictions that remain a challenge. Here, we address this challenge by taking advantage of the size control offered by microfluidics, in combination with the size reduction provided by cosolvent-infused PFC bubbles through both condensation and cosolvent dissolution. In this approach, PFC bubbles with a high percentage of cosolvent (in this study, diethyl ether, DEE) are produced using microfluidics at a temperature above the boiling point. After synthesis, these bubbles become much smaller through both condensation of the gas into liquid droplets and from dissolution of the DEE into the continuous phase. This approach demonstrates that monodisperse, cosolvent-incorporated PFC bubbles can directly form monodisperse PFC nanodroplets a factor of 24 times smaller than the precursor bubbles. We also demonstrate that these nanoscale droplets can be converted to echogenic microbubbles after exposure to ultrasound, showing that these PFC nanodroplets are viable for the in situ production of ultrasound contrast agents. We show that this system can overcome the minimum droplet size limit of standard microfluidics, and is a powerful new tool for generating monodisperse, PFC phase-change ultrasound contrast agents for treating and imaging cancer.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered