Merck
  • Monitoring of nitrite, nitrate, chloride and sulfate in environmental samples using electrophoresis microchips coupled with contactless conductivity detection.
Monitoring of nitrite, nitrate, chloride and sulfate in environmental samples using electrophoresis microchips coupled with contactless conductivity detection.
Talanta (2015-11-26)
Camilla Benevides Freitas, Roger Cardoso Moreira, Maria Gizelda de Oliveira Tavares, Wendell K T Coltro
ABSTRACT
This report describes the development of an analytical methodology on microchip electrophoresis (ME) devices coupled with capacitively coupled contactless conductivity detection (C(4)D) to monitor inorganic anions in environmental samples. The buffer composition as well as detection operating parameters were optimized to achieve the best separation selectivity and detector sensitivity, respectively. Electrophoretic separations of Cl(-), NO3(-), SO4(2-) and NO2(-) were successfully performed within 60s using a running buffer composed of 30mmol L(-1) latic acid and 15mmol L(-1)l-histidine (His). The best detectability levels were found applying a sinusoidal wave with 1100-kHz-frequency and 60-Vpp amplitude. Quantitative analyzes of inorganic anions were carried out in the presence of Cr2O7(2-) ion as internal standard (IS), which ensured great repeatability in terms of migration times (<1%) and peak areas (6.2-7.6%) for thirty consecutive injections. The analytical performance revealed a linear behavior for concentration ranges between 0-120μmol L(-1) (Cl(-), NO2(-) and NO3(-)) and 0-60μmol L(-1) (SO4(2-)) and limits of detection (LODs) varying from 2.0 to 4.9μmol L(-1). The concentration levels of anionic species were determined in aquarium, river and biofertilizer samples with recovery values between 91% and 105%. The nitrification steps associated with conversion of ammonium to nitrite followed by the conversion of nitrite to nitrate were successfully monitored in a simulated environment without fishes during a period of twelve weeks. Lastly, the monitoring of anionic species was carried out during eight weeks in an aquarium environment containing ten fishes from Danio rerio (Ciprynidae). The recorded data revealed the absence of nitrite and a gradual increase on the ammonium and nitrate concentration levels during eight weeks, thus suggesting the direct conversion of ammonium to nitrate. Based on the data herein reported, the proposed analytical methodology can be used for routine environmental analysis.
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
SAFC
L-Histidine
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)