Merck
  • Conjugated Polyelectrolyte-Induced Self-Assembly of Alkynylplatinum(II) 2,6-Bis(benzimidazol-2'-yl)pyridine Complexes.
Conjugated Polyelectrolyte-Induced Self-Assembly of Alkynylplatinum(II) 2,6-Bis(benzimidazol-2'-yl)pyridine Complexes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-10-03)
Kevin Chan, Clive Yik-Sham Chung, Vivian Wing-Wah Yam
ABSTRACT
Water-soluble cationic alkynylplatinum(II) 2,6-bis(benzimidazol-2'-yl)pyridine (bzimpy) complexes have been demonstrated to undergo supramolecular assembly with anionic polyelectrolytes in aqueous buffer solution. Metal-metal-to-ligand charge transfer (MMLCT) absorptions and triplet MMLCT ((3) MMLCT) emissions have been found in UV/Vis absorption and emission spectra of the electrostatic assembly of the complexes with non-conjugated polyelectrolytes, driven by Pt⋅⋅⋅Pt and π-π interactions among the complex molecules. Interestingly, the two-component ensemble formed by [Pt(bzimpy-Et){CCC6 H4 (CH2 NMe3 -4)}]Cl2 (1) with para-linked conjugated polyelectrolyte (CPE), PPE-SO3 (-) , shows significantly different photophysical properties from that of the ensemble formed by 1 with meta-linked CPE, mPPE-Ala. The helical conformation of mPPE-Ala allows the formation of strong mPPE-Ala-1 aggregates with Pt⋅⋅⋅Pt, electrostatic, and π-π interactions, as revealed by the large Stern-Volmer constant at low concentrations of 1. Together with the reasonably large Förster radius, large HOMO-LUMO gap and high triplet state energy of mPPE-Ala to minimize both photo-induced charge transfer (PCT) and Dexter triplet energy back-transfer (TEBT) quenching of the emission of 1, efficient Förster resonance energy transfer (FRET) from mPPE-Ala to aggregated 1 molecules and strong (3) MMLCT emission have been found, while the less strong PPE-SO3 (-) -1 aggregates and probably more efficient PCT and Dexter TEBT quenching would account for the lack of (3) MMLCT emission in the PPE-SO3 (-) -1 ensemble.
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Bromoethane, ReagentPlus®, ≥99%
Sigma-Aldrich
Bromoethane, reagent grade, 98%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Potassium bromide, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium 4-vinylbenzenesulfonate, technical, ≥90% (T)
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 98%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloroplatinate(II), ≥99.9% trace metals basis