Merck
  • In vivo study of immunogenicity and kinetic characteristics of a quantum dot-labelled baculovirus.
In vivo study of immunogenicity and kinetic characteristics of a quantum dot-labelled baculovirus.
Biomaterials (2015-06-29)
Meng Wang, Zhenhua Zheng, Jin Meng, Han Wang, Man He, Fuxian Zhang, Yan Liu, Bin Hu, Zike He, Qinxue Hu, Hanzhong Wang
ABSTRACT
Nanomaterials conjugated with biomacromolecules, including viruses, have great potential for in vivo applications. Therefore, it is important to evaluate the safety of nanoparticle-conjugated macromolecule biomaterials (Nano-mbio). Although a number of studies have assessed the risks of nanoparticles and macromolecule biomaterials in living bodies, only a few of them investigated Nano-mbios. Here we evaluated the in vivo safety profile of a quantum dot-conjugated baculovirus (Bq), a promising new Nano-mbio, in mice. Each animal was injected twice intraperitoneally with 50 μg virus protein labelled with around 3*10(-5)nmol conjugated qds. Control animals were injected with PBS, quantum dots, baculovirus, or a mixture of quantum dots and baculovirus. Blood, tissues and body weight were analysed at a series of time points following both the first and the second injections. It turned out that the appearance and behaviour of the mice injected with Bq were similar to those injected with baculovirus alone. However, combination of baculovirus and quantum dot (conjugated or simply mixed) significantly induced stronger adaptive immune responses, and lead to a faster accumulation and longer existence of Cd in the kidneys. Thus, despite the fact that both quantum dot and baculovirus have been claimed to be safe in vivo, applications of Bq in vivo should be cautious. To our knowledge, this is the first study examining the interaction between a nanoparticle-conjugated virus and a living body from a safety perspective, providing a basis for in vivo application of other Nano-mbios.
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Supelco
N,N′-Bis(acryloyl)cystamine, BioReagent, suitable for electrophoresis