Merck
  • Polysorbate 20 Degradation in Biopharmaceutical Formulations: Quantification of Free Fatty Acids, Characterization of Particulates, and Insights into the Degradation Mechanism.
Polysorbate 20 Degradation in Biopharmaceutical Formulations: Quantification of Free Fatty Acids, Characterization of Particulates, and Insights into the Degradation Mechanism.
Molecular pharmaceutics (2015-10-01)
Anthony Tomlinson, Barthélemy Demeule, Baiwei Lin, Sandeep Yadav
ABSTRACT
Polysorbate 20 (PS20), a commonly used surfactant in biopharmaceuticals, showed degradation upon long-term (∼18-36 months) storage of two monoclonal antibody (mAb, mAb-A, and mAb-B) drug products at 2-8 °C. The PS20 degradation resulted in the accumulation of free fatty acids (FFA), which ultimately precipitated to form particles upon long-term storage. This study documents the development, qualification, and application of a method for FFA quantification in soluble and insoluble fraction of protein formulation. The method was applied to the quantification of capric acid, lauric acid, myristic acid, palmitic/oleic acid, and stearic acid in placebo as well as active protein formulations on stability. Quantification of FFA in both the soluble and insoluble fraction of mAb-A and mAb-B provided a better mechanistic understanding of PS20 degradation and the dynamics of subsequent fatty acid particle formation. Additionally, the use of this method for monitoring and quantitation of the FFA on real time storage stability appears to aid in identifying batches with higher probability for particulate formation upon extended storage at 5 °C.
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%