Merck
Gestational Diabetes Impairs Human Fetal Postprandial Brain Activity.
The Journal of clinical endocrinology and metabolism (2015-10-16)
Katarzyna Linder, Franziska Schleger, Isabelle Kiefer-Schmidt, Louise Fritsche, Stefanie Kümmel, Martin Heni, Magdalene Weiss, Hans-Ulrich Häring, Hubert Preissl, Andreas Fritsche
ABSTRACT
Gestational diabetes (GDM) influences the fetal phenotype. In the present study, our aim was to determine the effect of GDM specifically on fetal brain activity. Pregnant participants underwent an oral glucose tolerance test (OGTT, 75 g). At 0, 60, and 120 minutes, maternal metabolism was determined, and fetal auditory evoked fields were recorded with a fetal magnetoencephalographic device. All measurements were performed at the fMEG Center in Tübingen. Twelve women with GDM and 28 normal glucose-tolerant (NGT) pregnant women participated on a voluntary basis. OGTT (75 g, 120 minutes) was used in this study. Fetal auditory evoked response latencies were determined for this study. In the fetuses of NGT women, latencies decreased between 0 and 60 minutes from 260 ± 90 to 206 ± 74 ms (P = .008) and remained stable until 120 minutes (206 ± 74 vs 230 ± 79, P =.129). In fetuses of women with GDM, there was no change in response latencies during OGTT (P = .11). Sixty minutes after glucose ingestion, fetal latencies in the GDM group were longer than in the NGT group (296 ± 82 vs 206 ± 74 ms, P = .001). Linear regression revealed a significant effect of maternal glucose, insulin levels, and insulin sensitivity on response latencies after 60 minutes. Fetal postprandial brain responses were slower in the offspring of women with GDM. This might indicate that gestational diabetes directly affects fetal brain development and may lead to central nervous insulin resistance in the fetus.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Glucose solution, BioUltra, Molecular Biology, ~20% in H2O
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture