Merck
  • Combined evaluation of LC3B puncta and HMGB1 expression predicts residual risk of relapse after adjuvant chemotherapy in breast cancer.
Combined evaluation of LC3B puncta and HMGB1 expression predicts residual risk of relapse after adjuvant chemotherapy in breast cancer.
Autophagy (2015-10-29)
Sylvain Ladoire, Frédérique Penault-Llorca, Laura Senovilla, Cécile Dalban, David Enot, Clara Locher, Nicole Prada, Vichnou Poirier-Colame, Kariman Chaba, Laurent Arnould, François Ghiringhelli, Pierre Fumoleau, Marc Spielmann, Suzette Delaloge, Marie Laure Poillot, Patrick Arveux, Aicha Goubar, Fabrice Andre, Laurence Zitvogel, Guido Kroemer
ABSTRACT
In spite of adjuvant chemotherapy, a significant fraction of patients with localized breast cancer (BC) relapse after optimal treatment. We determined the occurrence of cytoplasmic MAP1LC3B/LC3B (microtubule-associated protein 1 light chain 3B)-positive puncta, as well as the presence of nuclear HMGB1 (high mobility group box 1) in cancer cells within surgical BC specimens by immunohistochemistry, first in a test cohort (152 patients) and then in a validation cohort of localized BC patients who all received adjuvant anthracycline-based chemotherapy (1646 patients). Cytoplasmic LC3B(+) puncta inversely correlated with the intensity of SQSTM1 staining, suggesting that a high percentage cells of LC3B(+) puncta reflects increased autophagic flux. After setting optimal thresholds in the test cohort, cytoplasmic LC3B(+) puncta and nuclear HMGB1 were scored as positive in 27.2% and 28.6% of the tumors, respectively, in the validation cohort, while 8.7% were considered as double positive. LC3B(+) puncta or HMGB1 expression alone did not constitute independent prognostic factors for metastasis-free survival (MFS) in multivariate analyses. However, the combined positivity for LC3B(+) puncta and nuclear HMGB1 constituted an independent prognostic factor significantly associated with prolonged MFS (hazard ratio: 0.49 95% confidence interval [0.26-0.89]; P = 0.02), and improved breast cancer specific survival (hazard ratio: 0.21 95% confidence interval [0.05-0.85]; P = 0.029). Subgroup analyses revealed that within patients with poor-prognosis BC, HMGB1(+) LC3B(+) double-positive tumors had a better prognosis than BC that lacked one or both of these markers. Altogether, these results suggest that the combined positivity for LC3B(+) puncta and nuclear HMGB1 is a positive predictor for longer BC survival.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution