Merck
  • Respiratory syncytial virus nonstructural proteins 1 and 2 are crucial pathogenic factors that modulate interferon signaling and Treg cell distribution in mice.
Respiratory syncytial virus nonstructural proteins 1 and 2 are crucial pathogenic factors that modulate interferon signaling and Treg cell distribution in mice.
Virology (2015-08-25)
Pu Yang, Junwen Zheng, Shouyi Wang, Pin Liu, Meng Xie, Dongchi Zhao
ABSTRACT
Respiratory syncytial virus (RSV) nonstructural (NS) proteins 1 and 2 have multiple functions in suppressing the innate immune response and modulating T helper cell subset differentiation. However, little is known about the roles of NS proteins as independent virulence factors. We investigated the effects of recombinant NS1- and NS2-expressing plasmids on the pathogenesis of murine respiratory tissues and splenetic Foxp3+ regulatory T (Treg) cell distribution. Both NS proteins caused weight loss in mice, and NS2 transfection resulted in a persistent weight loss. NS1 dramatically suppressed the induction of interferon beta and interferon-induced GTP-binding protein Mx1. NS1 and NS2 demonstrated different effects in regulating Treg cell differentiation; NS2 increased the proportion of Tregs, whereas NS1 suppressed it. Inhibiting either NS1 or NS2 alleviated the pathology of lung tissues. Thus, NS1 and NS2 are independent pathogenic factors and could be targets for therapeutic strategies in treating RSV infection.
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
SAFC
Sodium deoxycholate
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)