Merck
  • Recombinant production and evaluation of an antibacterial L-amino acid oxidase derived from flounder Platichthys stellatus.
Recombinant production and evaluation of an antibacterial L-amino acid oxidase derived from flounder Platichthys stellatus.
Applied microbiology and biotechnology (2015-02-11)
Kosuke Kasai, Kenro Hashiguchi, Hiroki Takahashi, Ayano Kasai, Sadanori Takeda, Manabu Nakano, Takashi Ishikawa, Toshiya Nakamura, Tomisato Miura
ABSTRACT
Fish produce mucus substances as a defensive outer barrier against several bacterial infections. We have recently identified an antibacterial L-amino acid oxidase (psLAAO1) in the mucus layer of the flounder Platichthys stellate. In this study, the antibacterial protein psLAAO1 was expressed as a secretory bioactive recombinant protein in the methylotrophic yeast Pichia pastoris. The recombinant psLAAO1 inhibited the growth of bacteria to the same levels as native psLAAO1 present in mucus. In particular, Staphylococci and Yersinia were strongly suppressed, showing the highest growth retardation of the 21 species and strains tested. Moreover, Staphylococcus epidermidis was most sensitive to psLAAO1 with a minimum inhibitory concentration (MIC) of 0.078 μg/mL, whereas Escherichia coli was essentially resistant to psLAAO1 with a MIC of >10 μg/mL. Interestingly, psLAAO1-treated E. coli were found to upregulate the expression of the btuE gene, which encodes glutathione peroxidase (GPx). The biochemical function of GPx is to reduce free hydrogen peroxide and is induced under response to reactive oxygen species (ROS). Thus, E. coli confers resistance to the reduced free hydrogen peroxide produced by psLAAO1 by increasing GPx levels. Furthermore, the growth of Staphylococcus aureus was completely inhibited in the presence of recombinant psLAAO1. The morphology of psLAAO1-treated S. aureus showed cell surface damage, the formation of large aggregates and the cells showed severe deformations. Western blot analysis showed that psLAAO1 binds to the surface of S. aureus. Therefore, psLAAO1 binds to the surface of LAAO-sensitive S. aureus and directs peroxidative activity at the surface of the bacterial membrane.
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, Molecular Biology, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
1,4-Dithioerythritol, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
SAFC
Iodoacetamide
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, BioUltra