Merck
  • Distinct Plasma Bile Acid Profiles of Biliary Atresia and Neonatal Hepatitis Syndrome.
Distinct Plasma Bile Acid Profiles of Biliary Atresia and Neonatal Hepatitis Syndrome.
Journal of proteome research (2015-10-10)
Kejun Zhou, Jun Wang, Guoxiang Xie, Ying Zhou, Weihui Yan, Weihua Pan, Yanran Che, Ting Zhang, Linda Wong, Sandi Kwee, Yongtao Xiao, Jie Wen, Wei Cai, Wei Jia
ABSTRACT
Biliary atresia (BA) is a severe chronic cholestasis disorder of infants that leads to death if not treated on time. Neonatal hepatitis syndrome (NHS) is another leading cause of neonatal cholestasis confounding the diagnosis of BA. Recent studies indicate that altered bile acid metabolism is closely associated with liver injury and cholestasis. In this study, we systematically measured the bile acid metabolome in plasma of BA, NHS, and healthy controls. Liver bile acids were also measured using biopsy samples from 48 BA and 16 NHS infants undergoing operative cholangiography as well as 5 normal adjacent nontumor liver tissues taken from hepatoblastoma patients as controls. Both BA and NHS samples had significantly elevated bile acid levels in plasma compared to normal controls. BA patients showed a distinct bile acid profile characterized by the higher taurochenodeoxycholic acid (TCDCA) level and lower chenodeoxycholic acid (CDCA) level than those in NHS patients. The ratio of TCDCA to CDCA in plasma was significantly higher in BA compared to healthy infants (p < 0.001) or NHS (p < 0.001). The area under receiver operating characteristic curve for TCDCA/CDCA to differentiate BA from NHS was 0.923 (95% CI: 0.862-0.984). These findings were supported by significantly altered expression levels of bile acid transporters and nuclear receptors in liver including farnesoid X receptor (FXR), small heterodimer partner (SHP), bile salt export pump (BSEP), and multidrug resistant protein 3 (MDR3) in BA compared to NHS. Taken together, the plasma bile acid profiles are distinct in BA, NHS, and normal infants, as characterized by the ratio of TCDCA/CDCA differentially distributed among the three groups of infants.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%