Merck
  • β-Cyclodextrin-Propyl Sulfonic Acid Catalysed One-Pot Synthesis of 1,2,4,5-Tetrasubstituted Imidazoles as Local Anesthetic Agents.
β-Cyclodextrin-Propyl Sulfonic Acid Catalysed One-Pot Synthesis of 1,2,4,5-Tetrasubstituted Imidazoles as Local Anesthetic Agents.
Molecules (Basel, Switzerland) (2015-11-17)
Yan Ran, Ming Li, Zong-Ze Zhang
ABSTRACT
Some functionalized 1,2,4,5-tetrasubstituted imidazole derivatives were synthesized using a one-pot, four component reaction involving 1,2-diketones, aryl aldehydes, ammonium acetate and substituted aromatic amines. The synthesis has been efficiently carried out in a solvent free medium using β-cyclodextrin-propyl sulfonic acid as a catalyst to afford the target compounds in excellent yields. The local anesthetic effect of these derivatives was assessed in comparison to lidocaine as a standard using a rabbit corneal and mouse tail anesthesia model. The three most potent promising compounds were subjected to a rat sciatic nerve block assay where they showed considerable local anesthetic activity, along with minimal toxicity. Among the tested analogues, 4-(1-benzyl-4,5-diphenyl-1H-imidazol-2-yl)-N,N-dimethylaniline (5g) was identified as most potent analogue with minimal toxicity. It was further characterized by a more favourable therapeutic index than the standard.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis