Merck
  • Molecular marker approaches for tracking redox damage and protection in keratins.
Molecular marker approaches for tracking redox damage and protection in keratins.
Journal of cosmetic science (2014-03-08)
Jolon M Dyer, Charisa D Cornellison, Anita J Grosvenor, Stefan Clerens, Santanu Deb-Choudhury
ABSTRACT
There is increasing awareness of the importance of reductive and oxidative (redox) protein damage in protein-based materials including, hair, wool, nails, and skin. Light-induced damage to protein-based materials is of particular concern because of its impact on age-related degradation and product life spans. Consequently, cosmetic applications frequently target hair and skin restoration, where the integrity of the constituent filamentous proteins is essential to a healthy appearance. The keratins constitute an important subset of the structural proteins within skin, hair, and wool. We will introduce a means to assess damage to this important group of proteins at the molecular level, utilizing proteomic techniques to track the formation or degradation of sensitive peptides within intermediate filament proteins. The degradation of three molecular markers of redox damage, the peptides SFGYR, LASDDFR, and DVEEWYIR, along with the formation of their oxidized products, is demonstrated after exposure to ultraviolet A, ultraviolet B, and blue light. The method is shown to be suitable for evaluating the protective effect of treatments, as lower levels of oxidative markers were observed after the application of a protective fiber treatment. Molecular-level redox tracking will allow more targeted design and evaluation of protection and repair treatments for protein systems.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder