Merck
  • Electrospray droplet impact secondary ion mass spectrometry using a vacuum electrospray source.
Electrospray droplet impact secondary ion mass spectrometry using a vacuum electrospray source.
Rapid communications in mass spectrometry : RCM (2015-07-07)
R Takaishi, Y Sakai, K Hiraoka, H Wada, S Morita, T Nakashima, H Nonami
ABSTRACT
In electrospray droplet impact (EDI) developed in our laboratory, an atmospheric pressure electrospray source has been used. To increase the ion beam intensity and reduce the evacuation load, a vacuum electrospray cluster ion source using a silica capillary was developed. A silica capillary with a tip inner diameter of 8 µm was used for vacuum electrospray using aqueous 10% methanol. To stabilize the flow rate of the liquid for nano-electrospray, a home-made constant pressure liquid pump was also developed. By using the silica tip nano-electrospray emitter and a constant pressure pump, stable electrospray with flow rate of 22 nL/min was realized without using any heating system such as laser irradiation. Comparative study of mass spectra obtained by atmospheric pressure EDI (A-EDI) and vacuum EDI (V-EDI) was made for various samples such as thermometer molecule, peptide, polystyrene, Alq(3), NPD, C(60), indium, and SiO(2). V-EDI showed slightly milder ionization than A-EDI. Because V-EDI gave higher target current (5-10 nA) than A-EDI (a few nA at most), V-EDI secondary ion mass spectrometry (SIMS) would be a useful technique for the surface and interface analysis.
Sigma-Aldrich
N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine, 96%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine, sublimed grade, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Benzidine, ≥98%