Merck
  • Unusual clear cell, lymphoplasmacyte-rich, dural-based tumor with divergent differentiation: a tricky case mimicking a meningioma.
Unusual clear cell, lymphoplasmacyte-rich, dural-based tumor with divergent differentiation: a tricky case mimicking a meningioma.
Human pathology (2015-04-29)
Clelia Miracco, Marzia Toscano, Marie Aimée Gloria Munezero Butorano, Gennaro Baldino, Damiana Tacchini, Aurora Barone, Alfonso Cerase
ABSTRACT
We describe an unusual case of a recurrent dural neoplasm, previously diagnosed as meningioma. Histopathologically, the tumor is characterized by aggregates of divergently differentiated clear cells embedded in an abundant lymphoplasmacyte-rich stroma, mimicking a lymphoplasmacyte-rich meningioma. This study focuses on the histologic and immunohistochemical characterization of a unique dural-based tumor and provides useful guidelines for differentiating meningioma from other uncommon dural-based neoplasms. We propose that this recurrent dural neoplasm is a distinctive entity and, therefore, enlarges the spectrum of dural-based neoplasms that enter the differential diagnosis with meningiomas. Awareness of this tumor entity could prove useful for appropriate patient management.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast