Merck
  • Orally administrated dipeptide Ser-Tyr efficiently stimulates noradrenergic turnover in the mouse brain.
Orally administrated dipeptide Ser-Tyr efficiently stimulates noradrenergic turnover in the mouse brain.
Bioscience, biotechnology, and biochemistry (2015-05-23)
Takashi Ichinose, Kazuki Moriyasu, Akane Nakahata, Mitsuru Tanaka, Toshiro Matsui, Shigeki Furuya
ABSTRACT
In this study, we examined the effect of orally administrated dipeptides containing Tyr (Y) on the metabolism of catecholamines in mouse brains. We found that among eight synthetic dipeptides whose sequences are present frequently in soy proteins, Ser-Tyr (SY), Ile-Tyr, and Tyr-Pro had the highest apparent permeability coefficients in monolayers of human intestinal epithelial Caco-2 cells. When administrated orally, SY markedly increased tyrosine content in the cerebral cortex compared to the vehicle control, Ile-Tyr, Tyr-Pro, and Y alone. The oral administration of SY more effectively increased 3-methoxy-4-hydroxyphenylethyleneglycol, the principal metabolite of noradrenaline, in the cerebral cortex and hippocampus than did Ile-Tyr, Tyr-Pro, or Y alone. Central noradrenergic turnover was also markedly stimulated by SY administration. These in vivo observations strongly suggest that SY is more potent in boosting central catecholamine transmission, particularly the noradrenergic system, than Y alone or other dipeptides that include Y.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Tantalum(V) ethoxide, packaged for use in deposition systems
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
3-Methoxytyramine hydrochloride, 99% (AT)
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
3-Methoxytyramine hydrochloride, ≥95.5%, crystalline