Merck
  • Surface characterization and protein interaction of a series of model poly[acrylonitrile-co-(N-vinyl pyrrolidone)] nanocarriers for drug targeting.
Surface characterization and protein interaction of a series of model poly[acrylonitrile-co-(N-vinyl pyrrolidone)] nanocarriers for drug targeting.
International journal of pharmaceutics (2015-03-10)
Sven Staufenbiel, Marian Merino, Wenzhong Li, Mao-Dong Huang, Stefan Baudis, Andreas Lendlein, Rainer H Müller, Christian Wischke
ABSTRACT
The surface properties of intravenously injected nanoparticles determine the acquired blood protein adsorption pattern and subsequently the organ distribution and cellular recognition. A series of poly[acrylonitrile-co-(N-vinyl pyrrolidone)] (PANcoNVP) model nanoparticles (133-181 nm) was synthesized, in which the surface properties were altered by changing the molar content of NVP (0-33.8 mol%) as the more hydrophilic repeating unit. The extent of achieved surface property variation was comprehensively characterized. The residual sodium dodecyl sulfate (SDS) content from the synthesis was in the range 0.3-1.6 μgml(-1), potentially contributing to the surface properties. Surface hydrophobicity was determined by Rose Bengal dye adsorption, hydrophobic interaction chromatography (HIC) and aqueous two-phase partitioning (TPP). Particle charge was quantified by zeta potential (ZP) measurements including ZP-pH profiles. The interaction with proteins was analyzed by ZP measurements in serum and by adsorption studies with single proteins. Compared to hydrophobic polystyrene model nanoparticles, all PANcoNVP particles were very hydrophilic. Differences in surface hydrophobicity could be detected, which did not linearly correlate with the systematically altered bulk composition of the PANcoNVP nanoparticles. This proves the high importance of a thorough surface characterization applying a full spectrum of methods, complementing predictions solely based on bulk polymer composition.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis