Merck
  • Organochlorine pesticides and parasites in Mugil incilis collected in Cartagena Bay, Colombia.
Organochlorine pesticides and parasites in Mugil incilis collected in Cartagena Bay, Colombia.
Environmental science and pollution research international (2015-07-15)
Beatriz E Jaramillo-Colorado, Bárbara Arroyo-Salgado, Luis Carlos Ruiz-Garcés
ABSTRACT
Nematode parasites of the Anisakides family are often found in people living in countries where fish is consumed raw or partially cooked. This research shows the histological changes in the liver and spleen of Mugil incilis, collected in Cartagena Bay. These changes are associated with pollution by organochlorine pesticides and their possible influence on the parasite. Organochlorine compounds were extracted using the headspace-solid-phase microextraction (HS-SPME) technique. Residual amounts in the muscle of M. incilis such as β-HCH, γ-HCH, heptachlor, aldrin, endosulfan, 4,4'-DDE, and dieldrin, among others, were identified by gas chromatography connected to an electron capture detector, indicating that the fauna of Cartagena Bay are exposed to these pollutants. Histological analysis was carried out on liver and spleen samples of M. incilis which were fixed, processed, and embedded in paraffin. The presence of melano-macrophages, granulomes, and trematodes in the liver was the most important changes observed. Larval prevalence for the Anisakis spp. was determined to be 1.6%; for Pseudoterranova spp., 25.3%, and for Contracaecum spp., 57.8%. Other parasites such as acanthocephalans were also reported for a total of 15.3%. Nevertheless, no significant correlation between parasites and organochlorines was found. This study is the first to correlate the presence of organochlorine compounds and histological damage in the liver and spleen of M. incilis, with the presence of parasites in fish from Cartagena Bay (Colombia).
Product Description
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
SPME Portable Field Sampler, coating PDMS
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture