Merck
  • Complex autoregulation of the post-transcriptional regulator RsmA in Pseudomonas aeruginosa.
Complex autoregulation of the post-transcriptional regulator RsmA in Pseudomonas aeruginosa.
Microbiology (Reading, England) (2015-08-25)
Fabrice Jean-Pierre, Jonathan Perreault, Eric Déziel
ABSTRACT
RsmA is a post-transcriptional RNA-binding protein that acts as a pleiotropic global regulator of mRNAs in the opportunistic pathogen Pseudomonas aeruginosa. Upon binding to its target, RsmA impedes the translation of the mRNA by the ribosome. The RsmA regulon affects over 500 genes, many of which have been identified as important in the pathogenicity of P. aeruginosa. Whilst the regulatory function of RsmA is relatively well characterized, the genetic regulation of rsmA itself at the transcriptional and translational levels remains poorly understood. Here, we show that RsmA is capable of self-regulation through an unorthodox mechanism. This regulation occurs via direct interaction of the protein with an RsmA-binding site located in the early portion of its coding sequence. To the best of our knowledge this is the first report of such an unusual regulation in pseudomonads.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
2-Nitrophenyl β-D-galactopyranoside, ≥98% (enzymatic)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Spermidine, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O