Merck
  • Therapeutic effects of Lactobacillus casei Qian treatment in activated carbon-induced constipated mice.
Therapeutic effects of Lactobacillus casei Qian treatment in activated carbon-induced constipated mice.
Molecular medicine reports (2015-05-09)
Xin Zhao, Hua-Yi Suo, Yu Qian, Gui-Jie Li, Zhen-Hu Liu, Jian Li
ABSTRACT
In the present study, the therapeutic effects of Lactobacillus casei Qian (LC-Qian), the key microorganism in Tibetan yak yoghurt, on activated carbon-induced constipation were determined in vivo. ICR mice were treated with LC-Qian for nine days by oral administration. The body weight, defecation status, gastrointestinal transit and defecation time of mice were assessed, and the serum levels of motilin (MTL), gastrin (Gas), endothelin (ET), somatostatin (SS), acetylcholinesterase (AChE), substance P (SP) and vasoactive intestinal peptide (VIP) were further evaluated. Bisacodyl was used as the positive control. The time until the first black stool defecation following carbon intake of the normal, control, 100 mg/kg bisacodyl-treated, Lactobacillus bulgaricus (LB)-treated, LC-Qian (L)-and LC-Qian (H)-treated mice was 93, 231, 121, 194, 172 and 157 min, respectively. Following treatment with LC-Qian, the gastrointestinal transit was increased to 52.4% [LC-Qian (L)] and 65.8% [LC-Qian (H)], while that in the group treated with the common lactic acid bacteria of LB was 40.3%. The MTL, Gas, ET, AChE, SP and VIP serum levels were significantly increased and levels of SS were reduced in mice following LC-Qian treatment compared with those in the control mice (P<0.05). Reverse transcription quantitative polymerase chain reaction indicated that LC-Qian raised the c-Kit, GDNF as well as SCF mRNA expression levels and reduced the TRPV1 and NOS expression levels in tissue of the small intestine in mice. These results suggested that lactic acid bacteria prevent constipation in mice, among which LC-Qian was the most effective.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)