Merck
  • Waterproof Alkyl Phosphate Coated Fluoride Phosphors for Optoelectronic Materials.
Waterproof Alkyl Phosphate Coated Fluoride Phosphors for Optoelectronic Materials.
Angewandte Chemie (International ed. in English) (2015-07-28)
Hoang-Duy Nguyen, Chun Che Lin, Ru-Shi Liu
ABSTRACT
A facile approach for coating red fluoride phosphors with a moisture-resistant alkyl phosphate layer with a thickness of 50-100 nm is reported. K2 SiF6 :Mn(4+) particles were prepared by co-precipitation and then coated by esterification of P2 O5 with alcohols (methanol, ethanol, and isopropanol). This route was adopted to encapsulate the prepared phosphors using transition-metal ions as cross-linkers between the alkyl phosphate moieties. The coated phosphor particles exhibited a high water tolerance and retained approximately 87 % of their initial external quantum efficiency after aging under high-humidity (85 %) and high-temperature (85 °C) conditions for one month. Warm white-light-emitting diodes that consisted of blue InGaN chips, the prepared K2 SiF6 :Mn(4+) phosphors, and either yellow Y3 Al5 O12 :Ce(3+) phosphors or green β-SiAlON: Eu(2+) phosphors showed excellent color rendition.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Potassium permanganate, meets USP testing specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Potassium fluoride, powder, ≥99.9% trace metals basis