Merck
  • Uses of 1-(3-Cyano-4,5,6,7-tetrahydrobenzo[b]-thiophen-2-yl)-3-dodecanoylthiourea as a Building Block in the Synthesis of Fused Pyrimidine and Thiazine Systems.
Uses of 1-(3-Cyano-4,5,6,7-tetrahydrobenzo[b]-thiophen-2-yl)-3-dodecanoylthiourea as a Building Block in the Synthesis of Fused Pyrimidine and Thiazine Systems.
Chemical & pharmaceutical bulletin (2015-06-02)
Magdy Mohamed Hemdan, Heba Kamal Abd El-Mawgoude
ABSTRACT
The reaction of lauroyl isothiocyanate and 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile was used to synthesize the title compound 2. Compound 2 could serve as the main building block in the synthesis of many target heterocyclic systems. Various fused pyrimidines were synthesized in the reactions of compound 2 with sodium ethoxide, hydrazine hydrate, phenyl hydrazine, ethyl carbazate, thiourea, and/or 2-aminothiophenol. The structures of the synthesized compounds were confirmed by microanalytical and spectral data.
Sigma-Aldrich
Oxamide, 98%
Sigma-Aldrich
Thiophene, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%