Merck
  • Multifunctional gold nanoparticles as signal transducers for fabrication of 1:2 molecular demultiplexer.
Multifunctional gold nanoparticles as signal transducers for fabrication of 1:2 molecular demultiplexer.
Analytical and bioanalytical chemistry (2015-06-25)
Yi He, Haili Yu
ABSTRACT
A label-free and enzyme-free demultiplexer system for the fabrication of 1:2 molecular demultiplexer with luminol functionalized gold nanoparticles (Lum-AuNPs) as signal transducers was developed for the first time. The Lum-AuNPs had both chemiluminescence (CL) activity and surface plasmon resonance property. It was found that organothiols (RSH) could easily induce the aggregation of AuNPs via strong Au-S covalent interactions in the absence of hydrogen peroxide (H2O2), generating a red shift in the absorption band of AuNPs. However, the presence of H2O2 would readily oxidize RSH to disulfide (RS-SR), and the aggregation of Lum-AuNPs did not occur due to lack of the sulfhydryl group. Meanwhile, H2O2 could react with Lum-AuNPs, producing a strong CL emission owing to the enhancement effect of RSH on AuNPs-luminol-H2O2 CL system. Thus, RSH, H2O2, absorbance ratio, and CL intensity served as the signal input, address input, and two different signal outputs of the 1:2 molecular demultiplexer, respectively.
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Cysteamine, ≥98.0% (RT)
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%