Merck
  • Studies on antioxidative and immunostimulating fucogalactan of the edible mushroom Macrolepiota dolichaula.
Studies on antioxidative and immunostimulating fucogalactan of the edible mushroom Macrolepiota dolichaula.
Carbohydrate research (2015-06-16)
Surajit Samanta, Ashis K Nandi, Ipsita K Sen, Prasenjit Maity, Manabendra Pattanayak, K Sanjana P Devi, Somanjana Khatua, Tapas K Maiti, Krishnendu Acharya, Syed S Islam
ABSTRACT
A water soluble fucogalactan (PS-II) of an average molecular weight ∼1.2×10(5) Da was isolated from the aqueous extract of an edible mushroom Macrolepiota dolichaula. It was composed of fucose, galactose and 3-O-methyl galactose in a molar ratio of nearly 1:4:1. Structural characterization of PS-II was carried out using total hydrolysis, methylation analysis, Smith degradation, and 1D/2D NMR experiments. These results indicated that the proposed repeating unit of the PS-II had a backbone chain consisting of four (1→6)- linked α-d-Galp residues, one residue methylated at O-3, and another one substituted at O-2 by (1→2)-α-d-Galp residue, which is terminated with a α-l-Fucp moiety. The PS-II exhibited the antioxidant properties in different in vitro test systems, and also showed in vitro macrophage activation in RAW 264.7 cell line as well as splenocyte and thymocyte activation in mouse cell culture medium.
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by phenol extraction
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D