Merck
  • Oxytetracycline, tetracycline, chlortetracycline and doxycycline in pasteurised cow's milk commercialised in Brazil.
Oxytetracycline, tetracycline, chlortetracycline and doxycycline in pasteurised cow's milk commercialised in Brazil.
Food additives & contaminants. Part B, Surveillance (2014-09-24)
Carolina Kato Prado, Flávio Dias Ferreira, Erika Bando, Miguel Machinski
ABSTRACT
The aim of this study was to investigate the presence of tetracycline residues in pasteurised cow's milk using high-performance liquid chromatography coupled with UV/VIS detection to determine the exposure of Brazilian's population to antibiotic residues. One hundred samples collected from the State of Paraná, Brazil, were analysed. Three of these samples were contaminated at the following concentrations: 121.8 µg·kg(-1) for oxytetracycline, 93.5 µg·kg(-1) for tetracycline and 134.6 µg·kg(-1) for chlortetracycline (61.6 µg·kg(-1)) and doxycycline (73.0 µg·kg(-1)). The median tetracycline residue concentration found in the samples was 42.3 µg·kg(-1), and the estimated daily intake (EDI) was 0.05 µg Kg(-1) bw day(-1) in Brazil. These results demonstrate that the occurrence of tetracycline in Brazilian milk was low (3%) and only for 2% above the maximum residue limit, so the risk to the population from the presence of these residues in milk was low (<1% of the acceptable daily intake).
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%