Merck
  • Pharmacokinetic interactions of OBE001 and betamethasone in healthy female volunteers.
Pharmacokinetic interactions of OBE001 and betamethasone in healthy female volunteers.
Journal of clinical pharmacy and therapeutics (2015-04-23)
O Pohl, M-C Homery, F Lemaux, A Patat, A Chollet
ABSTRACT
To treat preterm labour, antenatal corticosteroids and tocolytics are often co-administered. OBE001 is an orally active oxytocin receptor antagonist under development for preterm labour treatment. Co-administration of OBE001 and betamethasone to determine pharmacokinetic interactions was studied during an open-label, randomized, three-period crossover study. Twelve healthy post-menopausal volunteers received either two consecutive OBE001 administrations of 600 mg/day, two intramuscular injections of 12 mg/day betamethasone or the two drugs administered in combination. The area under the plasma concentration-time curve (AUC), the maximum plasma concentration (Cmax) and the time to Cmax (tmax) for OBE001 and betamethasone were measured. There was no effect on geometric mean Cmax after the second administration and AUCs of OBE001 [geometric mean ratio point estimate (90% CI): Cmax (Day2) 1·05 (0·98-1·12) and AUC(0-24 h )1·11 (0·99-1·23)/AUC(24 h-∞) 0·99 (0·93-1·06), respectively]; Cmax after the first administration together with betamethasone was increased by 12% [geometric mean ratio point estimates (90% CI): Cmax (Day1) 1·12 (0·96-1·32)]. Tmax after concomitant administration with betamethasone occurred with a median delay of 1 h. Geometric mean Cmax and AUCs of betamethasone were not affected by concomitant OBE001 administration [geometric mean ratio point estimate (90% CI): Cmax (Day1) 1·02 (0·98-1·07)/Cmax (Day2) 1·03 (0·98-1·08) and AUC(0-24 h) 1·07 (1·04-1·11)/AUC(24 h-∞ )1·04 (1·01-1·08), respectively], with no effect on median tmax . No subject was discontinued from the study due to adverse events. AUC and Cmax of the betamethasone and OBE001 combination did not differ significantly between treatments. Co-administration of OBE001 and betamethasone was well tolerated and resulted in a tmax median delay of 1 h for OBE001 but not for betamethasone. Co-administration of OBE001 and betamethasone in clinics is feasible and does not require any specific precaution or administration adaptation.
Sigma-Aldrich
Betamethasone, ≥98%