Merck
  • Amperometric magnetoimmunosensor for ErbB2 breast cancer biomarker determination in human serum, cell lysates and intact breast cancer cells.
Amperometric magnetoimmunosensor for ErbB2 breast cancer biomarker determination in human serum, cell lysates and intact breast cancer cells.
Biosensors & bioelectronics (2015-03-21)
U Eletxigerra, J Martinez-Perdiguero, S Merino, R Barderas, R M Torrente-Rodríguez, R Villalonga, J M Pingarrón, S Campuzano
ABSTRACT
A highly sensitive amperometric magnetoimmunosensor for the determination of ErbB2 protein, a well-known biomarker related to high-impact high-incidence diseases such as breast cancer, is described. A sandwich format involving the covalent immobilization of a specific capture antibody onto magnetic beads (MBs) and incubation of the modified MBs with a mixture solution of the antigen and a HRP-labeled detector antibody was used. The resulting modified MBs were captured on the surface of a disposable screen-printed carbon electrode (SPCE) and the amperometric responses at -0.20 V were measured. This ErbB2 magnetoimmunosensor exhibited a very low detection limit of 26 pg mL(-1) far below the established cut-off for this biomarker (15 ng mL(-1)) and was successfully applied to the quantitation of ErbB2 in human serum and cell lysates samples without any matrix effect. In addition, the developed assay allowed the assessment of ErbB2 status directly in intact breast cancer cells. The results correlated well with those obtained with a commercial ELISA method, thus demonstrating that the new magnetoimmunosensing platform offers a truthful and useful analytical tool to be easily applied in breast cancer diagnosis through either ErbB2 protein determination or breast cancer cell status detection.
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethanolamine hydrochloride, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Ethanolamine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%