Merck
  • Bone regeneration and gene expression in bone defects under healthy and osteoporotic bone conditions using two commercially available bone graft substitutes.
Bone regeneration and gene expression in bone defects under healthy and osteoporotic bone conditions using two commercially available bone graft substitutes.
Biomedical materials (Bristol, England) (2015-05-09)
Claire I A van Houdt, Carla R Tim, Murilo C Crovace, Edgar D Zanotto, Oscar Peitl, Dietmar J O Ulrich, John A Jansen, Nivaldo A Parizotto, Ana C Renno, Jeroen J J P van den Beucken
ABSTRACT
Biosilicate(®) and Bio-Oss(®) are two commercially available bone substitutes, however, little is known regarding their efficacy in osteoporotic conditions. The purpose of this study was to evaluate the osteogenic properties of both materials, at tissue and molecular level. Thirty-six Wistar rats were submitted to ovariectomy (OVX) for inducing osteoporotic conditions and sham surgery (SHAM) as a control. Bone defects were created in both femurs, which were filled with Biosilicate(®) or Bio-Oss(®), and empty defects were used as control. For the healthy condition both Biosilicate(®) and Bio-Oss(®) did not improve bone formation after 4 weeks. Histomorphometric evaluation of osteoporotic bone defects with bone substitutes showed more bone formation, significant for Bio-Oss(®). Molecular biological evaluation was performed by gene-expression analysis (Runx-2, ALP, OC, OPG, RANKL). The relative gene expression was increased with Biosilicate(®) for all genes in OVX rats and for Runx-2, ALP, OC and RANKL in SHAM rats. In contrast, with Bio-Oss(®), the relative gene expression of OVX rats was similar for all three groups. For SHAM rats it was increased for Runx-2, ALP, OC and RANKL. Since both materials improved bone regeneration in osteoporotic conditions, our results suggest that bone defects in osteoporotic conditions can be efficiently treated with these two bone substitutes.
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
DBP, 98% (HPLC)
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)