Merck
  • Determination of free fatty acids in human serum by HPLC with fluorescence detection.
Determination of free fatty acids in human serum by HPLC with fluorescence detection.
Journal of chromatographic science (2014-07-19)
Minami Nishikiori, Hideaki Iizuka, Hideaki Ichiba, Kiyomi Sadamoto, Takeshi Fukushima
ABSTRACT
It has been suggested that serum concentrations of polyunsaturated essential fatty acids correlate with the symptoms or severity of various diseases, including depression and Alzheimer-type dementia, and that determination of serum fatty acids might be important for disease diagnosis. Thus, we developed to analyze serum fatty acids in healthy individuals by using a high-performance liquid chromatography method with fluorescence detection, because free fatty acids have a carboxyl group that can be derivatized with a fluorescent reagent, 4-N,N-dimethylaminosulfonyl-7-N-(2-aminoethyl)amino-2,1,3-benzoxadiazole. This approach could quantify five types of free fatty acids [α-linolenic acid (ALA), palmitoleic acid (PLA), arachidonic acid (AA), linoleic acid (LA) and oleic acid (OA)] in human serum. The detection limits of the method were in the range of 2.29-4.75 fmol (signal-to-noise ratio 3), and absolute concentrations of ALA, PLA, AA, LA and OA were 8.27 ± 1.04, 18.8 ± 2.95, 49.9 ± 4.03, 230 ± 18.1 and 201 ± 22.1 µM, respectively.
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥98%
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Heptadecanoic acid, ≥98%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)