Merck
  • Endoscopy-Assisted Total Mastectomy Followed by Immediate Pedicled Transverse Rectus Abdominis Musculocutaneous (TRAM) Flap Reconstruction: Preliminary Results of 48 Patients.
Endoscopy-Assisted Total Mastectomy Followed by Immediate Pedicled Transverse Rectus Abdominis Musculocutaneous (TRAM) Flap Reconstruction: Preliminary Results of 48 Patients.
Surgical innovation (2014-08-15)
Hung-Wen Lai, Hurng-Sheng Wu, Kun-Lin Chuang, Dar-Ren Chen, Tsai-Wang Chang, Shou-Jen Kuo, Shou-Tung Chen, Yao-Lung Kuo
ABSTRACT
Endoscopy-assisted breast surgery performed through minimal axillary and/or periareolar incisions is a viable option for patients with breast cancer. In this study, we report the preliminary results of patients with breast cancer who underwent endoscopy-assisted total mastectomy (EATM) followed immediately by pedicled transverse abdominis musculocutaneous (TRAM) flap reconstruction. Patients in this study comprised women with breast cancer who received EATM and pedicled TRAM flap reconstruction. Clinicopathologic characteristics, type of surgery, complications, and rate of recurrence were recorded. The cosmetic outcomes were evaluated objectively by the surgeons and subjectively by the patients at 3-month postoperative follow-up. A total of 48 patients underwent 49 EATM procedures followed by pedicled TRAM flap reconstruction. Of them, 79.6% underwent endoscopic-assisted nipple-sparing mastectomy and 20.4% received endoscopic-assisted skin-sparing mastectomy. The types of cancer among these patients included ductal carcinoma in situ in 34.7%, stage I cancer in 36.7%, stage II cancer in 24.5%, and stage IIIa cancer in 4.1% patients. Mean tumor size was 2.1 ± 1.4 cm. There were no cases of flap failure. Partial nipple areolar complex ischemia/necrosis occurred in 4 (10.3%) patients; however, all cases resolved after conservative treatment. In the aesthetic outcome evaluation, EATM + TRAM were associated with 89.8% good, 8.2% fair, and 2% unsatisfactory result. No local recurrence was observed during the follow-up period. EATM followed immediately by pedicled TRAM flap reconstruction is a safe procedure and results in good cosmetic outcome in women with early-stage breast cancer.
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution