Merck
  • Effect of Qing'e formula on the in vitro differentiation of bone marrow-derived mesenchymal stem cells from proximal femurs of postmenopausal osteoporotic mice.
Effect of Qing'e formula on the in vitro differentiation of bone marrow-derived mesenchymal stem cells from proximal femurs of postmenopausal osteoporotic mice.
BMC complementary and alternative medicine (2015-07-25)
Bo Shuai, Lin Shen, Rui Zhu, Piqi Zhou
ABSTRACT
Qing'e formula (QEF), prepared from an ancient Chinese recipe, was previously suggested to regulate bone metabolism and improve bone mineral density in patients with osteoporosis. To study the effects of medicated serum containing QEF on the in vitro differentiation of bone marrow-derived mesenchymal stem cells (BMSCs) isolated from the proximal femurs of postmenopausal osteoporosis (PMOP) mice. Using an established mouse model of PMOP, mononuclear cells were isolated from the bone marrow present in the proximal femurs and cultured. PMOP mice were also randomly divided into four groups: the untreated group (Group A) and the groups treated with respectively low (Group B), medium (Group C), and high (Group D) concentrations of QEF. Serum was isolated from each and used to treat the cultured BMSCs in conjunction with recombinant human bone morphogenetic protein-2 (rhBMP-2). Cell morphology, proliferation rates, intracellular alkaline phosphatase (ALP) activity, and transforming growth factor-beta 1 (TGF-β1) mRNA expression were evaluated. QEF-treated serum, particularly that containing moderate and high concentrations, appears to enhance the rhBMP-2-mediated changes in cell morphology, proliferation, and differentiation (determined via the expression of TGF-β1 mRNA and ALP activity) observed in the BMSCs isolated from PMOP mice. QEF may play a role in the prevention and treatment of PMOP by enhancing the activity of rhBMP-2.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, Carrier Free, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, BMP-2, recombinant, expressed in HEK 293 cells, HumanKine, suitable for cell culture
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C