Merck
  • Hexafluoroisopropanol-modified cetyltrimethylammonium bromide/sodium dodecyl sulfate vesicles as a pseudostationary phase in electrokinetic chromatography.
Hexafluoroisopropanol-modified cetyltrimethylammonium bromide/sodium dodecyl sulfate vesicles as a pseudostationary phase in electrokinetic chromatography.
Journal of chromatography. A (2015-06-06)
Yu Tian, Yunfang Li, Jie Mei, Bin Deng, Yuxiu Xiao
ABSTRACT
A novel catanionic surfactant vesicle system, formulated from hexafluoroisopropanol (HFIP), cetyltrimethylammonium bromide (CTAB) and sodium dodecyl sulfate (SDS), was developed as pseudostationary phase (PSP) in electrokinetic chromatography (EKC). HFIP, as an organic modifier with the prominent properties of ionization, hydrogen bond donor and hydrophobicity, was used to effectively promote the spontaneous vesicle formation from CTAB/SDS mixed aqueous solutions, where precipitates are easy to occur due to long carbon chains, and adjust the performance of CTAB/SDS vesicles. The physical features (size and viscosity) and electrophoretic parameters (electroosmotic mobility, electrophoretic mobility and elution range) of HFIP-modified CTAB/SDS vesicles were characterized as HFIP volume content (0-4%, v/v), CTAB/SDS molar ratio (2:8-7:3mol/mol) and total surfactant concentration (10-50mM) varying, respectively. The 3% v/v HFIP-modified CTAB/SDS (3:7mol/mol, 50mM) vesicle system proves to have the largest mean diameter (288.20nm) and the widest elution range (12.41), which is also much wider than that of the corresponding other four PSP systems including trifluoroethanol (TFE)-modified CTAB/SDS vesicles (5.69), isopropanol-modified CTAB/SDS micelles (2.03), HFIP-modified SDS micelles (4.86) and unmodified SDS micelles (3.12). The chromatographic performance of the HFIP-modified vesicle system was evaluated by separating eight polycyclic aromatic hydrocarbons, nitrotoluene positional isomers, five positively charged and five negatively charged/neutral drugs, respectively. Baseline or near-baseline separation was achieved for each series of solutes. Compared with the TFE-modified vesicle system, as well as the HFIP-modified and unmodified SDS micelle systems, the HFIP-modified vesicle system shows the best separation selectivity, the highest or comparable efficiency, and the lowest retention.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
3-Nitrotoluene, 99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Phenanthrene, 98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O