Merck
  • Fibrous Scaffolds with Varied Fiber Chemistry and Growth Factor Delivery Promote Repair in a Porcine Cartilage Defect Model.
Fibrous Scaffolds with Varied Fiber Chemistry and Growth Factor Delivery Promote Repair in a Porcine Cartilage Defect Model.
Tissue engineering. Part A (2015-09-25)
Iris L Kim, Christian G Pfeifer, Matthew B Fisher, Vishal Saxena, Gregory R Meloni, Mi Y Kwon, Minwook Kim, David R Steinberg, Robert L Mauck, Jason A Burdick
ABSTRACT
Current clinically approved methods for cartilage repair are generally based on either endogenous cell recruitment (e.g., microfracture) or chondrocyte delivery (e.g., autologous chondrocyte implantation). However, both methods culminate in repair tissue with inferior mechanical properties and the addition of biomaterials to these clinical interventions may improve their efficacy. To this end, the objective of this study was to investigate the ability of multipolymer acellular fibrous scaffolds to improve cartilage repair when combined with microfracture in a large animal (i.e., minipig) model. Composite scaffolds were formulated from a combination of hyaluronic acid (HA) fibers and poly(ɛ-caprolactone) (PCL) fibers, either with or without transforming growth factor-β3 (TGFβ3). After 12 weeks in vivo, material choice and TGFβ3 delivery had a significant impact on outcomes; specifically, PCL scaffolds without TGFβ3 had inferior gross appearance and reduced mechanical properties, whereas HA scaffolds that released TGFβ3 resulted in improved histological scores and increased type 2 collagen content. Importantly, analysis of the overall dataset revealed that histology, but not gross appearance, was a better predictor of mechanical properties. This study highlights the importance of scaffold properties on in vivo cartilage repair as well as the need for numerous quantitative outcome measures to fully evaluate treatment methods.
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Rhodamine B, suitable for fluorescence
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
SAFC
L-Proline
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%