Merck
  • Toxicity profiling of several common RNAi-based nanomedicines: a comparative study.
Toxicity profiling of several common RNAi-based nanomedicines: a comparative study.
Drug delivery and translational research (2015-03-20)
Dalit Landesman-Milo, Dan Peer
ABSTRACT
RNAi-based nanomedicine platforms (RNPs) have progressed from tools to study gene expression in vitro into clinical trials. Numerous RNPs strategies have been documented with an efficient ability to condense RNAi payloads and induce potent gene silencing. Moreover, some of these RNPs have been explored in various animal models, and some have even made it to the clinic. Still, there is lack of a clinically approved RNAi-based delivery strategy most probably due to unpredicted clinical toxicity. In this study, we prepared common RNPs such as cationic liposomes, polyamines, and hyaluronan-coated lipid-based nanoparticles and tested these strategies for global toxicity parameters such as changes in bodyweight, liver enzyme release, and hematological profiling. We found that polyamines such as polyethyleneimine and Poly-L-lysine released high levels of liver enzymes into the serum and reduced C57BL/6 mice bodyweight upon three intravenous injections. In addition, these polyamines dramatically reduced the total number of leukocytes, suggesting an immune suppression mechanism, while cationic liposomes, which also increased liver enzymes levels in the serum, elevated the total number of leukocytes probably by activation of Toll-like receptors 2 and 4. Coating the liposomes with hyaluronan, a hydrophilic glycosaminoglycan, provided a protective layer and did not induce adverse effects upon multiple intravenous injections. These findings suggest that there is an urgent need to develop gold standards for nanotoxicity in the field of RNAi that will be embraced by the RNAi community.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications