Merck
  • Evaluation of new transparent tungsten containing nanocomposites for radiation protection screens.
Evaluation of new transparent tungsten containing nanocomposites for radiation protection screens.
Radiation protection dosimetry (2015-03-31)
Diana Adliene, Egidijus Griskonis, Neringa Vaiciunaite, Rita Plaipaite-Nalivaiko
ABSTRACT
Evaluation of the new lead-free optically transparent nanocomposites is provided in this paper along with the introduction to their fabrication technologies and the results of investigation of their X-ray attenuating and optical properties before and after their exposure to high doses. These materials might be considered for the construction of X-ray protective screens that are used to protect medical personnel against scattered X rays during close-to patient operations. Rough selection of possible composites was performed according to X-ray attenuation modelling results. Experimental nanocomposites were prepared dissolving tungstates in different solvents or embedding tungsten structures into polymer matrix. It was found that the lead equivalent thickness of fabricated experimental samples varied from 0.15 to 0.75 mmPb and was higher for samples, containing polyanions of phosphotungstic acid. Transparency of the experimental samples to visible light varied from 20 to 60 % and was slightly increasing with irradiation dose due to radiation-induced processes in polymers.
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Ethylene glycol 5 M solution