Merck
  • One-pot green synthesis of oxygen-rich nitrogen-doped graphene quantum dots and their potential application in pH-sensitive photoluminescence and detection of mercury(II) ions.
One-pot green synthesis of oxygen-rich nitrogen-doped graphene quantum dots and their potential application in pH-sensitive photoluminescence and detection of mercury(II) ions.
Talanta (2015-05-25)
Bingfang Shi, Liangliang Zhang, Chuanqing Lan, Jingjin Zhao, Yubin Su, Shulin Zhao
ABSTRACT
Nitrogen doping has been a powerful method to modulate the properties of carbon materials for various applications, and N-doped graphene quantum dots (GQDs) have gained remarkable interest because of their unique chemical, electronic, and optical properties. Herein, we introduce a facile one-pot solid-phase synthesis strategy for N-doped GQDs using citric acid (CA) as the carbon source and 3,4-dihydroxy-L-phenylalanine (L-DOPA) as the N source. The as-prepared N-GQDs with oxygen-rich functional groups are uniform with an average diameter of 12.5 nm. Because of the introduction of nitrogen atoms, N-GQDs exhibit excitation-wavelength-independent fluorescence with the maximum emission at 445 nm, and a high quantum yield of 18% is achieved at an excitation wavelength of 346 nm. Furthermore, a highly efficient fluorosensor based on the as-prepared N-GQDs was developed for the detection of Hg(2+) because of the effective quenching effect of metal ions via nonradiative electron transfer. This fluorosensor exhibits high sensitivity toward Hg(2+) with a detection limit of 8.6 nM. The selectivity experiments reveal that the fluorescent sensor is specific for Hg(2+). Most importantly, the practical use of the sensor based on N-GQDs for Hg(2+) detection was successfully demonstrated in river-water samples.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Silver nitrate, ≥99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh