Merck
  • Extracellular vimentin interacts with insulin-like growth factor 1 receptor to promote axonal growth.
Extracellular vimentin interacts with insulin-like growth factor 1 receptor to promote axonal growth.
Scientific reports (2015-07-15)
Michiko Shigyo, Tomoharu Kuboyama, Yusuke Sawai, Masahito Tada-Umezaki, Chihiro Tohda
ABSTRACT
Vimentin, an intermediate filament protein, is generally recognised as an intracellular protein. Previously, we reported that vimentin was secreted from astrocytes and promoted axonal growth. The effect of extracellular vimentin in neurons was a new finding, but its signalling pathway was unknown. In this study, we aimed to determine the signalling mechanism of extracellular vimentin that facilitates axonal growth. We first identified insulin-like growth factor 1 receptor (IGF1R) as a receptor that is highly phosphorylated by vimentin stimulation. IGF1R blockades diminished vimentin- or IGF1-induced axonal growth in cultured cortical neurons. IGF1, IGF2 and insulin were not detected in the neuron culture medium after vimentin treatment. The combined drug affinity responsive target stability method and western blotting analysis showed that vimentin and IGF1 interacted with IGF1R directly. In addition, immunoprecipitation and western blotting analyses confirmed that recombinant IGF1R bound to vimentin. The results of a molecular dynamics simulation revealed that C-terminal residues (residue number 330-407) in vimentin are the most appropriate binding sites with IGF1R. Thus, extracellular vimentin may be a novel ligand of IGF1R that promotes axonal growth in a similar manner to IGF1. Our results provide novel findings regarding the role of extracellular vimentin and IGF1R in axonal growth.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerin, meets USP testing specifications
Pricing and availability is not currently available.
SAFC
L-Glutamine
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine
Pricing and availability is not currently available.