Merck
Injectable sodium pentobarbital: Stability at room temperature.
Journal of pharmacological and toxicological methods (2015-08-04)
Sydney M Priest, Timothy P Geisbuhler
ABSTRACT
Sodium pentobarbital (Nembutal) is a barbiturate used in research as an anesthetic in many animal models. The injectable form of this drug has lately become difficult to procure and prohibitively expensive. Due to this lack of availability, researchers have begun to compound injectable sodium pentobarbital from so-called "nonpharmaceutical" pentobarbital. Some oversight agencies have objected to this practice, claiming a lack of quality control and degradation of the drug. We sought with this study to establish both: 1) a protocol for the preparation of injectable sodium pentobarbital, and 2) standard operating procedures to monitor the quality of the preparation and degradation of the drug over time. Our preparation consists of a mixture of sodium pentobarbital in alkaline aqueous solution, propylene glycol, and ethanol. Pentobarbital content in this preparation was assayed by high-pressure liquid chromatography (HPLC). We also assayed pentobarbital content over time in preparations of various ages up to 6 years old. We determined that the drug degraded at a maximum of 0.5% per year in our preparation (alkaline water/propylene glycol/ethanol) when stored in the dark at room temperature. A yellow discoloration developed after about 2 years, which we have arbitrarily determined disqualifies the preparation from use as an anesthetic. Attempts to spectroscopically assay this discoloration were not successful. Pentobarbital sodium (CID: 14075609).
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.