Merck
  • Occult hepatitis B virus infection among people with a family history of chronic hepatitis B virus infection.
Occult hepatitis B virus infection among people with a family history of chronic hepatitis B virus infection.
Journal of medical virology (2015-05-13)
Zhenhua Zhang, Ling Zhang, Yu Dai, Lei Jin, Binghu Sun, Qian Su, Xu Li
ABSTRACT
The prevalence of occult hepatitis B virus infection (OBI) among people with a family history of chronic hepatitis B virus (HBV) infection is unclear. Serum samples were collected from 747 hepatitis B surface antigen (HBsAg)-negative people with a family history of HBV infection and 579 HBsAg-negative volunteer blood donors. The presence of HBV DNA was evaluated using nested PCR with primers specific for the X, S, and C regions of HBV. The Pre-S1/Pre-S2/ S region PCR products for the OBI group and their family members with chronic HBV infection (control group) were sequenced and compared. The prevalence of OBI was 8.0% (60/747) among HBsAg-negative people with a family history of chronic HBV infection, compared to 2.6% (15/579) among the blood donors (P < 0.05). The prevalence of HBV genotype B infection was lower in the OBI group than in the control group (P = 0.031). The substitution rates in the major hydrophilic region and the "a" determinant seemed to be higher in the OBI group (0.893 vs. 0.507; 1.042 vs. 0.403, respectively), and stop codon mutations more frequent in the OBI sequences (OBI: 2/26, 7.7% vs. 0/31, 0%). However, none of these differences was statistically significant (P = 0.237, 0.199, 0.201, respectively). In summary, the prevalence of OBI among HBsAg-negative people with a family history of chronic HBV infection was significantly higher than that in Chinese blood donors. However, S region mutations and the escape mechanism are not likely to be the major causes of increased prevalence of OBI.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis